2-Nitrophenylboronic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dihydroxy(2-nitrophenyl)borane, (2-Nitrophenyl)boronic acid, 2-Nitrobenzeneboronic acid, (2-Nitrophenyl)metaboric acid
L17988-06
L17988-03
AAAL17988-06EA
282
USD
AAAL17988-06
AAAL17988-03
2-Nitrophenylboronic acid 96%
2-Nitrophenylboronic acid
Purity: 96%
CAS No.: 5570-19-4
Molecular Formula: C6H6BNO4
Formula Weight: 166.93
Melting Point: 149-151°
Beilstein Reference: 3032097
Formula:
C₆H₆BNO₄ Melting Pt: 149…151 °C Storage Temperature: Ambient |
MDL Number:
MFCD00161358 CAS Number: 5570-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...