2-Chloro-5-hydroxypyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-5-pyridinol, 6-Chloro-3-pyridinol

L17701-03 L17701-06
AAL17701-03EA 96.9 USD
AAL17701-03 AAL17701-06
2-Chloro-5-hydroxypyridine 97%
2-Chloro-5-hydroxypyridine

Purity: 97% CAS No.: 41288-96-4 Molecular Formula: C5H4ClNO Formula Weight: 129.55 Melting Point: 152-158° MDL No.: MFCD00234050

Formula: C₅H₄ClNO
MW: 129.55
Melting Pt: 152…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00234050
CAS Number: 41288-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR