1-Boc-indole 97%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
tert-Butyl 1-indolecarboxylate
L17601-03
L17601-06
AAAL17601-03EA
45.3
USD
AAAL17601-03
AAAL17601-06
1-Boc-indole 97%
1-Boc-indole
Purity: 97%
CAS No.: 75400-67-8
Molecular Formula: C13H15NO2
Formula Weight: 217.27
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5430
Boiling Point: 113-114°/0.2mm
Beilstein Reference: 4674239
MDL No.: MFCD02093939
Formula:
C₁₃H₁₅NO₂ MW: 217.27 g/mol Boiling Pt: 113…114 °C/0.2 mm Density: 1.070 Storage Temperature: Ambient |
MDL Number:
MFCD02093939 CAS Number: 75400-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...