5-Methyl-2-hexanone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Isoamyl methyl ketone, Methyl isoamyl ketone
L17584-0F
L17584-AP
L17584-AE
AAAL17584-0FEA
172
USD
AAAL17584-0F
AAAL17584-AP
AAAL17584-AE
5-Methyl-2-hexanone 99%
5-Methyl-2-hexanone
Purity: 99%
CAS No.: 110-12-3
Molecular Formula: C7H14O
Formula Weight: 114.19
Flash Point: 41°(105°F)
Refractive Index: 1.4075
Melting Point: -74°
Boiling Point: 144-145°
Beilstein Reference: 506163
UN No.: 2302
MDL No.: MFCD00008950
Formula:
C₇H₁₄O Boiling Pt: 144…145 °C Melting Pt: –74 °C Density: 0.81 g/cm³ (20 °C) Flash Pt: 41 °C (105 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008950 CAS Number: 110-12-3 EINECS: 203-737-8 UN: 2302 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...