5-Methylisoxazole-3-carboxylic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

L17577-06 L17577-14
AAAL17577-06EA 79.3 USD
AAAL17577-06 AAAL17577-14
5-Methylisoxazole-3-carboxylic acid 98+%
5-Methylisoxazole-3-carboxylic acid

Purity: 98+% CAS No.: 3405-77-4 Molecular Formula: C5H5NO3 Formula Weight: 127.10 Melting Point: 171-173° Beilstein Reference: 114094

Formula: C₅H₅NO₃
MW: 127.1 g/mol
Melting Pt: 171…173 °C
Storage Temperature: Ambient
MDL Number: MFCD01318162
CAS Number: 3405-77-4
EINECS: 222-289-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR