Dimethyl azelate 97%

Supplier: Thermo Scientific Chemicals

L17458-06 L17458-14 L17458-22
AAL17458-06EA 58.1 USD
AAL17458-06 AAL17458-14 AAL17458-22
Dimethyl azelate 97%
Dimethyl azelate

Purity: 97% CAS No.: 1732-10-1 Molecular Formula: C11H20O4 Formula Weight: 216.28 Flash Point: 110°(230°F) Refractive Index: 1.4370 Melting Point: 18° Boiling Point: 156°/20mm Beilstein Reference: 1710125 Merck Reference: 14,905 MDL No.: MFCD00025898

Formula: C₁₁H₂₀O₄
Boiling Pt: 156 °C/20 mm
Melting Pt: 18 °C
Density: 1.007
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00025898
CAS Number: 1732-10-1
EINECS: 217-060-0
Merck Index: 14,905

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR