2-Iodobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

L17446-03 L17446-06
AAAL17446-03EA 96.9 USD
AAAL17446-03 AAAL17446-06
2-Iodobenzaldehyde 98%
2-Iodobenzaldehyde

Purity: 98% CAS No.: 26260-02-6 Molecular Formula: C7H5IO Formula Weight: 232.02 Storage Temperature: 2°C to 8°C Melting Point: 36-37° Boiling Point: 102°/3mm Beilstein Reference: 1927296 MDL No.: MFCD00039570

Formula: C₇H₅IO
MW: 232.02 g/mol
Boiling Pt: 102 °C/3 mm
Melting Pt: 36…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00039570
CAS Number: 26260-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR