2,4,6,8-Tetramethylcyclotetrasiloxane 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16642-22
L16642-14
AAAL16642-22EA
491
USD
AAAL16642-22
AAAL16642-14
2,4,6,8-Tetramethylcyclotetrasiloxane 99%
2,4,6,8-Tetramethylcyclotetrasiloxane
Purity: 99%
CAS No.: 2370-88-9
Molecular Formula: C4H16O4Si4
Formula Weight: 240.51
Flash Point: 25°(77°F)
Refractive Index: 1.3870
Melting Point: -69°
Boiling Point: 134-135°
Beilstein Reference: 1787074
UN No.: 1993
MDL No.: MFCD00039567
Formula:
C₄H₁₆O₄Si₄ Boiling Pt: 134…135 °C Melting Pt: –69 °C Density: 0.99 g/cm³ (20 °C) Flash Pt: 25 °C (77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039567 CAS Number: 2370-88-9 EINECS: 219-137-4 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...