1,4-Dibutylbenzene 97+%
Supplier: Thermo Scientific Chemicals
L16396-03
L16396-09
AAL16396-03EA
41.4
USD
AAL16396-03
AAL16396-09
1,4-Dibutylbenzene 97+%
1,4-Dibutylbenzene
Purity: 97+%
CAS No.: 1571-86-4
Molecular Formula: C14H22
Formula Weight: 190.33
Flash Point: 110°(230°F)
Refractive Index: 1.4890
Melting Point: -31°
Boiling Point: 227°
Beilstein Reference: 1858055
MDL No.: MFCD01076591
Formula:
C₁₄H₂₂ MW: 190.33 Boiling Pt: 227 °C Melting Pt: –31 °C Density: 0.856 Flash Pt: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01076591 CAS Number: 1571-86-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...