3-(2-Aminoethyl)pyridine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-(Pyridin-3-yl)ethanamine, 2-(3-Pyridyl)ethylamine
L16311-06
L16311-03
AAAL16311-06EA
353
USD
AAAL16311-06
AAAL16311-03
3-(2-Aminoethyl)pyridine 98%
3-(2-Aminoethyl)pyridine
Purity: 98%
CAS No.: 20173-24-4
Molecular Formula: C7H10N2
Formula Weight: 122.17
Refractive Index: 1.5518
Odor: Amine-like
Boiling Point: 119-120°/19mm
Beilstein Reference: 111288
UN No.: 2735
MDL No.: MFCD00191604
Formula:
C₇H₁₀N₂ MW: 122.17 g/mol Boiling Pt: 54 °C (0.06 -rr) Storage Temperature: Ambient |
MDL Number:
MFCD00191604 CAS Number: 20173-24-4 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...