Chlorodimethylvinylsilane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dimethylvinylchlorosilane, Vinyldimethylchlorosilane
L16198-14
L16198-22
AAAL16198-14EA
65.26
USD
AAAL16198-14
AAAL16198-22
Chlorodimethylvinylsilane 97%
Chlorodimethylvinylsilane
Purity: 97%
CAS No.: 1719-58-0
Molecular Formula: C4H9ClSi
Formula Weight: 120.66
Flash Point: -12°(10°F)
Refractive Index: 1.4160
Boiling Point: 82-83°
Beilstein Reference: 1737688
UN No.: 2985
MDL No.: MFCD00018090
Formula:
C₄H₉ClSi MW: 120.65 g/mol Boiling Pt: 82…83 °C Density: 0.883 Flash Pt: -12 °C(10 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00018090 CAS Number: 1719-58-0 EINECS: 217-007-1 UN: 2985 ADR: "3",II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...