Methyl indole-4-carboxylate 99%

Supplier: Thermo Scientific Chemicals
Warning

L16021-03 L16021-06
AAAL16021-03EA 32.8 USD
AAAL16021-03 AAAL16021-MD AAAL16021-06
Methyl indole-4-carboxylate 99%
Methyl indole-4-carboxylate

Purity: 99% CAS No.: 39830-66-5 Molecular Formula: C10H9NO2 Formula Weight: 175.19 Melting Point: 68-71° Beilstein Reference: 141826

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00191222
CAS Number: 39830-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR