Methyl indole-6-carboxylate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 1H-indole-6-carboxylate
L16020-06
L16020-03
AAAL16020-06EA
68.6
USD
AAAL16020-06
AAAL16020-03
Methyl indole-6-carboxylate 98%
Methyl indole-6-carboxylate
Purity: 98%
CAS No.: 50820-65-0
Molecular Formula: C10H9NO2
Formula Weight: 175.19
Melting Point: 78-80°
Beilstein Reference: 141837
Formula:
C₁₀H₉NO₂ MW: 175.19 g/mol Melting Pt: 78…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00211063 CAS Number: 50820-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...