4-Amino-2-chlorobenzonitrile 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15924-14
L15924-22
L15924-06
AAAL15924-14EA
86.7
USD
AAAL15924-14
AAAL15924-22
AAAL15924-06
4-Amino-2-chlorobenzonitrile 98+%
4-Amino-2-chlorobenzonitrile
Purity: 98+%
CAS No.: 20925-27-3
Molecular Formula: C7H5ClN2
Formula Weight: 152.58
Melting Point: 116-119°
Beilstein Reference: 2936425
UN No.: 3439
MDL No.: MFCD00035926
Formula:
C₇H₅ClN₂ Boiling Pt: 301 °C (1013 hPa) Melting Pt: 162 - 163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035926 CAS Number: 20925-27-3 EINECS: 244-114-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...