(±)-Citronellal 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(+/-)-Citronellal, (±)-3,7-Dimethyloct-6-enal, (+/-)-3,7-Dimethyl-6-octenal
L15753-AE
L15753-AP
AAAL15753-AEEA
64.31
USD
AAAL15753-AE
AAAL15753-AP
(±)-Citronellal 96%
(±)-Citronellal
Purity: 96%
CAS No.: 106-23-0
Molecular Formula: C10H18O
Formula Weight: 154.25
Flash Point: 75°(167°F)
Refractive Index: 1.4460
Boiling Point: 206-208°
Beilstein Reference: 1720789
Merck Reference: 14,2329
MDL No.: MFCD00038090
Formula:
C₁₀H₁₈O MW: 154.25 g/mol Boiling Pt: 207 °C (1013 hPa) Melting Pt: -15 °C Density: 0.8548 g/cm³ (20 °C) Flash Pt: 75 °C(167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00038090 CAS Number: 106-23-0 EINECS: 203-376-6 Merck Index: 13,02353 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...