Methylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

L15589-03 L15589-06
AAAL15589-03EA 51.5 USD
AAAL15589-03 AAAL15589-06
Methylboronic acid 97%
Methylboronic acid

Purity: 97% CAS No.: 13061-96-6 Molecular Formula: CH5BO2 Formula Weight: 59.86 Odor: Odorless Melting Point: 89-94° Beilstein Reference: 1731087 MDL No.: MFCD00002105

Formula: CH₅BO₂
MW: 59.86 g/mol
Melting Pt: 89…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00002105
CAS Number: 13061-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR