1-Ethylpiperazine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L15539-36
L15539-22
AAAL15539-36EA
182
USD
AAAL15539-36
AAAL15539-22
1-Ethylpiperazine 98%
1-Ethylpiperazine
Purity: 98%
CAS No.: 5308-25-8
Molecular Formula: C6H14N2
Formula Weight: 114.19
Flash Point: 43°(109°F)
Refractive Index: 1.4690
Odor: Amine-like
Boiling Point: 157°
Beilstein Reference: 102971
UN No.: 2733
MDL No.: MFCD00059912
Formula:
C₆H₁₄N₂ Boiling Pt: 157 °C Melting Pt: -60 °C Density: 0.8989 g/cm³ (20 °C) Flash Pt: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00059912 CAS Number: 5308-25-8 EINECS: 226-166-8 UN: 2734 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...