3,4,5-Trimethoxyphenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,4,5-Trimethoxybenzeneboronic acid, Dihydroxy(3,4,5-trimethoxyphenyl)borane, (3,4,5-Trimethoxyphenyl)metaboric acid
L15191-06
L15191-03
AAAL15191-06EA
198
USD
AAAL15191-06
AAAL15191-03
3,4,5-Trimethoxyphenylboronic acid 98%
3,4,5-Trimethoxyphenylboronic acid
Purity: 98%
CAS No.: 182163-96-8
Molecular Formula: C9H13BO5
Formula Weight: 212.01
Melting Point: ca 245°
Beilstein Reference: 8205051
Formula:
C₉H₁₃BO₅ MW: 212.01 g/mol Melting Pt: ∼ 245 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075676 CAS Number: 182163-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...