Methacrylamide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15013-36
L15013-22
AAAL15013-36EA
55.68
USD
AAAL15013-36
AAAL15013-22
Methacrylamide 98%
Methacrylamide
Purity: 98%
CAS No.: 79-39-0
Molecular Formula: C4H7NO
Formula Weight: 85.11
Melting Point: 107-112°
Boiling Point: 215°
MDL No.: MFCD00008018
Formula:
C₄H₇NO MW: 85.11 g/mol Boiling Pt: 215 °C Melting Pt: 107…112 °C Density: 1.053 g/cm³ (20 °C) Flash Pt: 103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008018 CAS Number: 79-39-0 EINECS: 201-202-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...