3-Methylxanthine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14986-03
L14986-MD
AAAL14986-03EA
219
USD
AAAL14986-03
AAAL14986-MD
3-Methylxanthine 98+%
3-Methylxanthine
Purity: 98+%
CAS No.: 1076-22-8
Molecular Formula: C6H6N4O2
Formula Weight: 166.14
Melting Point:
Formula:
C₆H₆N₄O₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005580 CAS Number: 1076-22-8 EINECS: 214-058-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...