Boron tribromide 1 M in dichloromethane
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14880-AC
L14880-AE
AAAL14880-ACEA
63.18
USD
AAAL14880-AC
AAAL14880-AE
Boron tribromide 1 M in dichloromethane
Boron tribromide
CAS No.: 10294-33-4
Molecular Formula: BBr3
Formula Weight: 250.52
Refractive Index: 1.4340
Merck Reference: 14,1347
UN No.: 3390
MDL No.: MFCD00011312
Formula:
BBr₃ Density: 1.467 Storage Temperature: Ambient |
MDL Number:
MFCD00011312 CAS Number: 10294-33-4 EINECS: 233-657-9 UN: 3390 ADR: 6.1,I Merck Index: 14,1347 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...