1-Methyl-1H-pyrazole 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14831-06
L14831-14
AAAL14831-06EA
57
USD
AAAL14831-06
AAAL14831-14
1-Methyl-1H-pyrazole 97+%
1-Methyl-1H-pyrazole
Purity: 97+%
CAS No.: 930-36-9
Molecular Formula: C4H6N2
Formula Weight: 82.11
Flash Point: 35°(95°F)
Refractive Index: 1.4760
Odor: Pungent
Boiling Point: 127°
Beilstein Reference: 105207
UN No.: 1993
MDL No.: MFCD00144943
Formula:
C₄H₆N₂ Boiling Pt: 127 °C Density: 0.988 Flash Pt: 35 °C (95 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00144943 CAS Number: 930-36-9 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...