(±)-3-Buten-2-ol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (±)-Methyl vinyl carbinol, (±)-3-Hydroxy-1-butene, 1-Buten-3-ol

L14787-14 L14787-06
AAAL14787-14EA 114 USD
AAAL14787-14 AAAL14787-06
(±)-3-Buten-2-ol 97%
(±)-3-Buten-2-ol

Purity: 97% CAS No.: 598-32-3 Molecular Formula: C4H8O Formula Weight: 72.11 Flash Point: 20°(68°F) Refractive Index: 1.4140 Melting Point: -100° Boiling Point: 96-98° Beilstein Reference: 1361410 UN No.: 1986 MDL No.: MFCD00004543

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 96…98 °C
Melting Pt: < –100 °C
Density: 0.913 g/cm³ (20 °C)
Flash Pt: 20 °C(68 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004543
CAS Number: 598-32-3
EINECS: 209-929-8
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR