α,α,α,α',α',α'-Hexafluoro-2,5-xylidine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,5-Bis(trifluoromethyl)aniline

L14706-06 L14706-14
AAAL14706-06EA 119 USD
AAAL14706-06 AAAL14706-14
α,α,α,α',α',α'-Hexafluoro-2,5-xylidine 99%
α,α,α,α',α',α'-Hexafluoro-2,5-xylidine

Purity: 99% CAS No.: 328-93-8 Molecular Formula: C8H5F6N Formula Weight: 229.13 Flash Point: 71°(159°F) Refractive Index: 1.4338 Odor: Recognizable Boiling Point: 70-71°/15mm Beilstein Reference: 2653046 UN No.: 2810 MDL No.: MFCD00074940

Formula: C₈H₅F₆N
MW: 229.12 g/mol
Boiling Pt: 70…71 °C/15 mm
Density: 1.479 g/cm³ (20 °C)
Flash Pt: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00074940
CAS Number: 328-93-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR