1-Phenyl-2-pyrrolidone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Phenyl-2-pyrrolidinone

L14660-06 L14660-14 L14660-22
AAL14660-06EA 43 USD
AAL14660-06 AAL14660-14 AAL14660-22
1-Phenyl-2-pyrrolidone 99%
1-Phenyl-2-pyrrolidone

Purity: 99% CAS No.: 4641-57-0 Molecular Formula: C10H11NO Formula Weight: 161.20 Odor: Odorless Melting Point: 63-68° Boiling Point: 123°/0.2mm Beilstein Reference: 130685 MDL No.: MFCD00003192

Formula: C₁₀H₁₁NO
MW: 161.20 g/mol
Boiling Pt: 123°C/0.2mm
Melting Pt: 63…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00003192
CAS Number: 4641-57-0
EINECS: 225-069-6
ADR: 8,I
Merck Index: 13,01337

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR