Nujol (light oil) for IR spectrophotometry
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14480-AD
L14480-AP
AAAL14480-ADEA
40.83
USD
AAAL14480-AD
AAAL14480-AP
Nujol (light oil) for IR spectrophotometry
Nujol (light oil)
CAS No.: 8012-95-1
Flash Point: 220°(428°F)
Refractive Index: 1.4680
Melting Point: ca -20°
Boiling Point:
Boiling Pt:
300 - 500 °C (1013 hPa) Melting Pt: ∼ …20 °C Density: 0.86 g/cm³ (20 °C) Flash Pt: 215 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00131611 CAS Number: 8012-95-1 EINECS: 232-384-2 Merck Index: 13,07093 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...