Methyl acetate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 99%
CAS No.: 79-20-9
Molecular Formula: CH3COOCH3
Formula Weight: 74.08
Flash Point: -10°(14°F)
Refractive Index: 1.3620
Odor: Pleasant
Melting Point: -98°
Boiling Point: 56-58°
Beilstein Reference: 1736662
Merck Reference: 14,6008
UN No.: 1231
MDL No.: MFCD00008711
Formula:
C₃H₆O₂ Boiling Pt: 56…58 °C Melting Pt: –98 °C Density: 0.934 Flash Pt: –10 °C (14 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008711 CAS Number: 79-20-9 EINECS: 201-185-2 UN: 1231 ADR: 3,II Merck Index: 14,6008 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...