11-Bromo-1-undecanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 11-Bromoundecan-1-ol, 11-Bromoundecanol

L14448-06 L14448-14
AAAL14448-06EA 38.05 USD
AAAL14448-06 AAAL14448-14
11-Bromo-1-undecanol 97%
11-Bromo-1-undecanol

Purity: 97% CAS No.: 1611-56-9 Molecular Formula: Br(CH2)11OH Formula Weight: 251.21 Flash Point: 110°(230°F) Melting Point: 46-49° Boiling Point: 165-170°/1mm Beilstein Reference: 1741180 MDL No.: MFCD00004752

Formula: HO(CH₂)₁₀CH₂Br
MW: 251.21 g/mol
Boiling Pt: 165…170 °C/1 mm
Melting Pt: 46…49 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004752
CAS Number: 1611-56-9
EINECS: 216-554-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR