Diethyl sulfate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 98%
CAS No.: 64-67-5
Molecular Formula: (CH3CH2O)2SO2
Formula Weight: 154.19
Flash Point: 104°(219°F)
Refractive Index: 1.4005
Melting Point: -25°
Boiling Point: 208° dec.
Beilstein Reference: 1209714
Merck Reference: 14,3130
UN No.: 1594
MDL No.: MFCD00009099
Formula:
C₄H₁₀O₄S Boiling Pt: 208 °C (dec.) Melting Pt: –25 °C Density: 1.179 g/cm³ (20 °C) Flash Pt: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009099 CAS Number: 64-67-5 EINECS: 200-589-6 UN: 1594 ADR: 6.1,II Merck Index: 13,03156 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...