Fentin chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chlorotriphenylstannane, Triphenyltin chloride

L14289-14 L14289-36 L14289-22
AAAL14289-14EA 36.2 USD
AAAL14289-14 AAAL14289-36 AAAL14289-22
Fentin chloride 95%
Fentin chloride

Purity: 95% CAS No.: 639-58-7 Molecular Formula: C18H15ClSn Formula Weight: 385.48 Flash Point: 110°(230°F) Odor: Unpleasant Boiling Point: 240°/13.5mm Beilstein Reference: 524762 UN No.: 3146 MDL No.: MFCD00000519

Formula: C₁₈H₁₅ClSn
MW: 385.48 g/mol
Boiling Pt: 397
Melting Pt: 103…106 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000519
CAS Number: 639-58-7
EINECS: 211-358-4
UN: 3146
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR