Chrysin 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 5,7-Dihydroxy-2-phenyl-4H-chromen-4-one, 5,7-Dihydroxyflavone, 5,7-Dihydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 5,7-Dihydroxy-2-phenyl-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-phenyl-4-benzopyrone, 5,7-Dihydroxy-2-phenylchromen-4-one

L14178-06 L14178-14
AAAL14178-06EA 40.2 USD
AAAL14178-06 AAAL14178-14
Chrysin 98%
Chrysin

Purity: 98% CAS No.: 480-40-0 Molecular Formula: C15H10O4 Formula Weight: 254.24 Melting Point: 284-289° Beilstein Reference: 233276 Merck Reference: 14,2256 MDL No.: MFCD00006834

Formula: C₁₅H₁₀O₄
MW: 254.24 g/mol
Boiling Pt: 429
Melting Pt: 284…289 °C
Storage Temperature: Ambient
MDL Number: MFCD00006834
CAS Number: 480-40-0
EINECS: 207-549-7
Merck Index: 14,2256

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR