5-Bromopyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

L14172-14 L14172-22 L14172-06
AAAL14172-14EA 71.1 USD
AAAL14172-14 AAAL14172-22 AAAL14172-06
5-Bromopyrimidine 98%
5-Bromopyrimidine

Purity: 98% CAS No.: 4595-59-9 Molecular Formula: C4H3BrN2 Formula Weight: 158.99 Melting Point: 69-74° Beilstein Reference: 107326

Formula: C₄H₃BrN₂
MW: 158.99 g/mol
Boiling Pt: 168 °C (727 -rr)
Melting Pt: 75 - 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00006117
CAS Number: 4595-59-9
EINECS: 224-992-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR