3-Furonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Cyanofuran
L14019-06
L14019-03
AAAL14019-06EA
565
USD
AAAL14019-06
AAAL14019-03
AAAL14019-MD
3-Furonitrile 98%
3-Furonitrile
Purity: 98%
CAS No.: 30078-65-0
Molecular Formula: C5H3NO
Formula Weight: 93.09
Flash Point: 53°(127°F)
Refractive Index: 1.4790
Melting Point: 28-30°
Boiling Point: 150-151°
Beilstein Reference: 507009
UN No.: 2926
MDL No.: MFCD00067042
Formula:
C₅H₃NO MW: 93.08 g/mol Boiling Pt: 150…151 °C Melting Pt: 28…30 °C Flash Pt: 53 °C(127 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00067042 CAS Number: 30078-65-0 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...