4',7-Dimethoxyisoflavone 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 7-Methoxy-3-(4-methoxyphenyl)chromen-4-one, 7-Methoxy-3-(4-methoxyphenyl)-4-benzopyrone, 4',7-Dimethoxyisoflavone

L13838-MC L13838-04
AAAL13838-MCEA 44.4 USD
AAAL13838-MC AAL13838-ME AAAL13838-04
4',7-Dimethoxyisoflavone 97%
4',7-Dimethoxyisoflavone

Purity: 97% CAS No.: 1157-39-7 Molecular Formula: C17H14O4 Formula Weight: 282.30 Melting Point: 161-163° Beilstein Reference: 265550

Formula: C₁₇H₁₄O₄
MW: 282.3 g/mol
Melting Pt: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00075889
CAS Number: 1157-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR