D-(+)-Cystine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-(+)-Cystine, (H-D-Cys-OH)2, D-Cystine
L13772-03
L13772-06
AAL13772-03EA
99.8
USD
AAL13772-03
AAL13772-06
D-(+)-Cystine 98%
D-(+)-Cystine
Purity: 98%
CAS No.: 349-46-2
Molecular Formula: C6H12N2O4S2
Formula Weight: 240.30
Odor: Odorless
Melting Point: ca 250° dec.
Beilstein Reference: 1728093
Merck Reference: 14,2782
Optical Rotation: +219° (c=1 in 1N HCl)
MDL No.: MFCD00002610
Formula:
HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH MW: 240.3 g/mol Melting Pt: ∼ 250 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002610 CAS Number: 349-46-2 EINECS: 206-486-2 Merck Index: 13,02811 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...