2-Nitroimidazole 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Azomycin, 2-Nitro-1H-imidazole

L13546-MC L13546-ME
AAAL13546-MCEA 51.4 USD
AAAL13546-MC AAAL13546-ME
2-Nitroimidazole 98%
2-Nitroimidazole

Purity: 98% CAS No.: 527-73-1 Molecular Formula: C3H3N3O2 Formula Weight: 113.08 Melting Point: ca 287° dec. Beilstein Reference: 116444 Merck Reference: 14,921 UN No.: 2811 MDL No.: MFCD00005185

Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
Melting Pt: 279 - 284 °C
Storage Temperature: Ambient
MDL Number: MFCD00005185
CAS Number: 527-73-1
EINECS: 208-425-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,00923

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR