Potassium nitrite 97%

Supplier: Thermo Scientific Chemicals
Danger

L13289-22 L13289-36
AAAL13289-22EA 55.07 USD
AAAL13289-22 AAAL13289-36
Potassium nitrite 97%
Potassium nitrite

Purity: 97% CAS No.: 7758-09-0 Molecular Formula: KNO2 Formula Weight: 85.11 Odor: Odorless pH: 7.0-10.0 Melting Point: 350° dec. Merck Reference: 14,7649 UN No.: 1488 MDL No.: MFCD00011408

Formula: KNO₂
MW: 85.1 g/mol
Melting Pt: 387 °C
Density: 1.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011408
CAS Number: 7758-09-0
EINECS: 231-832-4
UN: 1488
ADR: 5.1,II
Merck Index: 13,07734

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR