3-Thiophenecarbonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Cyanothiophene, 3-Thienyl cyanide, Thiophene-3-carbonitrile

L13115-06 L13115-14
AAAL13115-06EA 61.6 USD
AAAL13115-06 AAAL13115-14
3-Thiophenecarbonitrile 98%
3-Thiophenecarbonitrile

Purity: 98% CAS No.: 1641-09-4 Molecular Formula: C5H3NS Formula Weight: 109.15 Flash Point: 86°(186°F) Refractive Index: 1.5615 Odor: Recognizable Boiling Point: 205° Beilstein Reference: 107104 UN No.: 3276 MDL No.: MFCD00151852

Formula: C₅H₃NS
MW: 109.15 g/mol
Boiling Pt: 205 °C
Density: 1.1956 g/cm³ (20 °C)
Flash Pt: 86 °C(186 °F)
Storage Temperature: Ambient
MDL Number: MFCD00151852
CAS Number: 1641-09-4
EINECS: 216-687-7
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR