Sodium ethoxide 21% (w/w) in ethanol

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium ethylate, Sodium ethanolate

L13083-0F L13083-AP L13083-AE
AAAL13083-0FEA 250.7 USD
AAAL13083-0F AAAL13083-AP AAAL13083-AE
Sodium ethoxide 21% (w/w) in ethanol
Sodium ethoxide

CAS No.: 141-52-6 Molecular Formula: C2H5NaO Formula Weight: 68.05 Flash Point: 20°(68°F) Refractive Index: 1.3875 Beilstein Reference: 3593646 Merck Reference: 14,8613 UN No.: 3274 MDL No.: MFCD00012417

Formula: C₂H₅NaO
MW: 68.05 g/mol
Density: 0.868
Flash Pt: 20 °C (68 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 2924
ADR: 3,III
Merck Index: 14,8613

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR