6-Bromo-1-hexene 95%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

L12909-06 L12909-03
AAAL12909-06EA 161 USD
AAAL12909-06 AAAL12909-03
6-Bromo-1-hexene 95%, Technical Grade
6-Bromo-1-hexene

Purity: 95% CAS No.: 2695-47-8 Molecular Formula: C6H11Br Formula Weight: 163.06 Flash Point: 54°(129°F) Refractive Index: 1.4652 Odor: Characteristic Boiling Point: 147° Beilstein Reference: 506203 UN No.: 1993 MDL No.: MFCD00000269

Formula: C₆H₁₁Br
MW: 163.06 g/mol
Boiling Pt: 147 °C
Density: 1.2169 g/cm³ (20 °C)
Flash Pt: 54 °C (129 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000269
CAS Number: 2695-47-8
EINECS: 220-267-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR