4-Bromo-1,1,2-trifluoro-1-butene 98%

Supplier: Thermo Scientific Chemicals
Danger

L12766-09 L12766-18
AAAL12766-09EA 175 USD
AAAL12766-09 AAAL12766-18
4-Bromo-1,1,2-trifluoro-1-butene 98%
4-Bromo-1,1,2-trifluoro-1-butene

Purity: 98% CAS No.: 10493-44-4 Molecular Formula: C4H4BrF3 Formula Weight: 188.98 Flash Point: 17°(62°F) Refractive Index: 1.4020 Odor: Recognizable Boiling Point: 95-98° Beilstein Reference: 1840755 UN No.: 1993 MDL No.: MFCD00039274

Formula: C₄H₄BrF₃
MW: 188.97 g/mol
Boiling Pt: 95 °C (1013 hPa)
Density: 1.639 g/cm³ (20 °C)
Flash Pt: 17 °C (62 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039274
CAS Number: 10493-44-4
EINECS: 234-019-2
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR