Indene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Purity: 97%
CAS No.: 95-13-6
Molecular Formula: C9H8
Formula Weight: 116.16
Storage Temperature: 2°C to 8°C
Flash Point: 58°(136°F)
Refractive Index: 1.5740
Odor: Aromatic
Melting Point: ca -2°
Boiling Point: 181-182°
Beilstein Reference: 635873
Merck Reference: 14,4939
UN No.: 3295
MDL No.: MFCD00003777
Formula:
C₉H₈ Boiling Pt: 181…182 °C Melting Pt: ca …2 °C Density: 0.998 g/cm³ (20 °C) Flash Pt: 58 °C(136 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003777 CAS Number: 95-13-6 EINECS: 202-393-6 UN: 3295 ADR: "3",III Merck Index: 13,04964 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...