4-(Trifluoromethyl)phenylhydrazine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-(4-(Trifluoromethyl)phenyl)hydrazine, 4-Hydrazinobenzotrifluoride
L12436-03
L12436-06
AAAL12436-03EA
40.2
USD
AAAL12436-03
AAAL12436-06
4-(Trifluoromethyl)phenylhydrazine 95%
4-(Trifluoromethyl)phenylhydrazine
Purity: 95%
CAS No.: 368-90-1
Molecular Formula: C7H7F3N2
Formula Weight: 176.14
Storage Temperature: 2°C to 8°C
Melting Point: 60-64°
Boiling Point: 119-121°/17mm
Beilstein Reference: 743759
MDL No.: MFCD00042508
Formula:
C₇H₇F₃N₂ Boiling Pt: 119…121 °C/17 mm Melting Pt: 60…64 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00042508 CAS Number: 368-90-1 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...