(S)-(+)-2-Octanol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
D(+)-2-Octanol, (S)-Octan-2-ol, (S)-(+)-Hexyl methyl carbinol, (S)-(+)-2-Hydroxy octane, (S)-(+)-Methyl hexyl carbinol
L12425-03
L12425-06
AAAL12425-03EA
38.6
USD
AAAL12425-03
AAAL12425-06
(S)-(+)-2-Octanol 99%
(S)-(+)-2-Octanol
Purity: 99%
CAS No.: 6169-06-8
Molecular Formula: C8H18O
Formula Weight: 130.23
Flash Point: 76°(169°F)
Refractive Index: 1.4260
Odor: Pleasant
Boiling Point: 179-180°
Beilstein Reference: 1719323
Merck Reference: 14,6752
Optical Rotation: +10° (c=1 in ethanol)
MDL No.: MFCD00064283
Formula:
C₈H₁₈O MW: 130.23 g/mol Boiling Pt: 179…180 °C Density: 0.822 g/cm³ (20 °C) Flash Pt: 76 °C (169 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064283 CAS Number: 6169-06-8 EINECS: 228-213-6 UN: 1986 ADR: 3,III Merck Index: 12,06850 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...