Sorbitan monolaurate

Supplier: Thermo Scientific Chemicals

Synonyms: Sorbitan monododecanoate, Sorbitan laurate, Span 20

L12099-30 L12099-0B
AAAL12099-30EA 58.32 USD
AAAL12099-30 AAAL12099-0B
Sorbitan monolaurate
Sorbitan monolaurate

CAS No.: 1338-39-2 Molecular Formula: C18H34O6 Formula Weight: 346.47 Flash Point: 110°(230°F) Refractive Index: 1.4740 Merck Reference: 14,8724 MDL No.: MFCD00005365

Formula: C₁₈H₃₄O₆
MW: 346.46 g/mol
Density: 1.032
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005365
CAS Number: 1338-39-2
EINECS: 215-663-3
Merck Index: 14,8724

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR