3-Methylheptane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12006-06
L12006-03
L12006-14
AAAL12006-06EA
128
USD
AAAL12006-06
AAL12006-03
AAL12006-14
3-Methylheptane 97%
3-Methylheptane
Purity: 97%
CAS No.: 589-81-1
Molecular Formula: C8H18
Formula Weight: 114.23
Flash Point: 7°(44°F)
Refractive Index: 1.3980
Melting Point: -121°
Boiling Point: 118-120°
Beilstein Reference: 1730777
UN No.: 1262
MDL No.: MFCD00027244
Formula:
C₈H₁₈ Boiling Pt: 118…120 °C Melting Pt: –121 °C Density: 0.710 Flash Pt: 7 °C(44 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00027244 CAS Number: 589-81-1 EINECS: 209-660-6 UN: 1262 ADR: "3",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...