4-Iodo-3-nitroanisole 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Iodo-4-methoxy-2-nitrobenzene
L11714-14
L11714-06
AAAL11714-14EA
239
USD
AAAL11714-14
AAAL11714-06
4-Iodo-3-nitroanisole 98+%
4-Iodo-3-nitroanisole
Purity: 98+%
CAS No.: 58755-70-7
Molecular Formula: C7H6INO3
Formula Weight: 279.04
Melting Point: 60-63°
Beilstein Reference: 2365592
Formula:
C₇H₆INO₃ Melting Pt: 60…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00024195 CAS Number: 58755-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...