1-Heptene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11487-AC
L11487-AE
AAAL11487-ACEA
103.35
USD
AAAL11487-AC
AAAL11487-AE
1-Heptene 98+%
1-Heptene
Purity: 98+%
CAS No.: 592-76-7
Molecular Formula: C7H14
Formula Weight: 98.19
Flash Point: -8°(17°F)
Refractive Index: 1.400
Melting Point: ca -119°
Boiling Point: 93-94°
Beilstein Reference: 1098332
UN No.: 2278
MDL No.: MFCD00009531
Formula:
C₇H₁₄ MW: 98.19 g/mol Boiling Pt: 93 °C (1013 hPa) Melting Pt: -119 °C Density: 0.7 g/cm³ (20 °C) Flash Pt: –8 °C (17 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009531 CAS Number: 592-76-7 EINECS: 209-767-8 UN: 2278 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...