3,5-Dihydroxybenzhydrazide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L11471-06
L11471-14
AAAL11471-06EA
73
USD
AAAL11471-06
AAAL11471-14
3,5-Dihydroxybenzhydrazide 98%
3,5-Dihydroxybenzhydrazide
Purity: 98%
CAS No.: 7732-32-3
Molecular Formula: C7H8N2O3
Formula Weight: 168.15
Melting Point: 256-257°
Beilstein Reference: 2642968
Formula:
C₇H₈N₂O₃ Melting Pt: 256…257 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017066 CAS Number: 7732-32-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...