2-Ethoxyphenyl isocyanate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Ethoxy-2-isocyanatobenzene

L11299-14 L11299-06
AAAL11299-14EA 131 USD
AAAL11299-14 AAAL11299-06
2-Ethoxyphenyl isocyanate 97%
2-Ethoxyphenyl isocyanate

Purity: 97% CAS No.: 5395-71-1 Molecular Formula: C9H9NO2 Formula Weight: 163.18 Storage Temperature: 2°C to 8°C Flash Point: 102°(215°F) Refractive Index: 1.5270 Boiling Point: 119-120°/20mm Beilstein Reference: 1102956 UN No.: 2206 MDL No.: MFCD00002007

Formula: C₉H₉NO₂
MW: 163.18 g/mol
Boiling Pt: 119…120 °C/20 mm
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 102 °C (215 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002007
CAS Number: 5395-71-1
UN: 2206
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR